Assay Details
Gene Symbol: csnk1db | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213250.1 | NP_998415.1 | 4 - 5 | 939 | 68 | |
GenBank mRNA | BC063953.1 | - | 4 - 5 | 939 | 68 | |
BC097045.1 | - | 4 - 5 | 972 | 68 | ||
BC165561.1 | - | 4 - 5 | 601 | 68 | ||
EU127825.1 | - | 4 - 5 | 567 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
