Assay Details
Gene Symbol: csnk1db | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213250.1 | NP_998415.1 | 8 - 9 | 1564 | 58 | |
GenBank mRNA | BC063953.1 | - | 8 - 9 | 1564 | 58 | |
BC097045.1 | - | 8 - 9 | 1597 | 58 | ||
BC165561.1 | - | 8 - 9 | 1226 | 58 | ||
EU127825.1 | - | 8 - 9 | 1192 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
