Assay Details
| Gene Symbol: eif5a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213262.1 | NP_998427.1 | 2 - 3 | 378 | 88 | |
| GenBank mRNA | BC048043.1 | - | 2 - 3 | 332 | 88 | |
| BC066558.1 | - | 2 - 3 | 378 | 88 | ||
| BC164906.1 | - | 1 - 2 | 205 | 88 | ||
More Information
Set Membership: |
|

eif5a2
Genomic Map