Assay Details
| Gene Symbol: eif5a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213262.1 | NP_998427.1 | 4 - 5 | 614 | 94 | |
| GenBank mRNA | BC048043.1 | - | 4 - 5 | 568 | 94 | |
| BC066558.1 | - | 4 - 5 | 614 | 94 | ||
| BC164906.1 | - | 3 - 4 | 441 | 94 | ||
More Information
Set Membership: |
|

eif5a2
Genomic Map