Assay Details
Gene Symbol: akirin2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213294.2 | NP_998459.2 | 1 - 2 | 483 | 60 | |
GenBank mRNA | AY394967.1 | - | 1 - 2 | 488 | 60 | |
BC045294.1 | - | 1 - 2 | 474 | 60 | ||
BC065319.1 | - | 1 - 2 | 412 | 60 | ||
BC097074.1 | - | 1 - 2 | 494 | 60 | ||
EH552945.1 | - | 1 - 2 | 495 | 60 | ||
EH584322.1 | - | 1 - 2 | 516 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
