Assay Details
| Gene Symbol: tmem59l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213337.2 | NP_998502.2 | 7 - 8 | 1011 | 64 | |
| GenBank mRNA | BC054584.1 | - | 7 - 8 | 957 | 64 | |
| BC154612.1 | - | 7 - 8 | 955 | 64 | ||
More Information
Set Membership: |
|

tmem59l
Genomic Map