Assay Details
Gene Symbol: lgals3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214693.2 | NP_999858.2 | 4 - 5 | 403 | 133 | |
GenBank mRNA | AY421705.1 | - | 4 - 5 | 392 | 133 | |
BC093113.1 | - | 4 - 5 | 413 | 133 | ||
BC107990.1 | - | 4 - 5 | 403 | 133 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
