Assay Details
| Gene Symbol: syncrip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214696.1 | NP_999861.1 | 7 - 8 | 916 | 63 | |
| XM_009294525.1 | XP_009292800.1 | 7 - 8 | 980 | 63 | ||
| XM_009294526.1 | XP_009292801.1 | 7 - 8 | 980 | 63 | ||
| XM_009294527.1 | XP_009292802.1 | 6 - 7 | 809 | 63 | ||
| GenBank mRNA | BC056750.1 | - | 7 - 8 | 916 | 63 | |
| BC068373.1 | - | 7 - 8 | 913 | 63 | ||
More Information
Set Membership: |
|

syncrip
Genomic Map