Assay Details
| Gene Symbol: bves | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001847.2 | NP_001001847.2 | 6 - 7 | 937 | 62 | |
| NM_001257162.1 | NP_001244091.1 | 6 - 7 | 910 | 62 | ||
| NM_001257163.1 | NP_001244092.1 | 6 - 7 | 937 | 62 | ||
| NM_001257164.1 | NP_001244093.1 | 6 - 7 | 910 | 62 | ||
| GenBank mRNA | AY293117.1 | - | 6 - 7 | 935 | 62 | |
| AY293118.1 | - | 6 - 7 | 908 | 62 | ||
| BC086956.1 | - | 6 - 7 | 910 | 62 | ||
| JN400527.1 | - | 6 - 7 | 935 | 62 | ||
More Information
Set Membership: |
|

bves
Genomic Map