Assay Details
| Gene Symbol: rps13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002079.1 | NP_001002079.1 | 4 - 5 | 335 | 76 | |
| GenBank mRNA | BC072552.1 | - | 4 - 5 | 335 | 76 | |
| BC163978.1 | - | 4 - 5 | 359 | 76 | ||
More Information
Set Membership: |
|

rps13
Genomic Map