Assay Details
| Gene Symbol: mrps15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002485.2 | NP_001002485.1 | 6 - 7 | 574 | 61 | |
| GenBank mRNA | BC076324.1 | - | 6 - 7 | 575 | 61 | |
More Information
Set Membership: |
|

mrps15
Genomic Map