Assay Details
Gene Symbol: rpl35a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002487.1 | NP_001002487.1 | 3 - 4 | 218 | 91 | |
GenBank mRNA | BC076321.1 | - | 3 - 4 | 218 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
