Assay Details
| Gene Symbol: aclya | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002649.2 | NP_001002649.2 | 10 - 11 | 1286 | 94 | |
| GenBank mRNA | BC076484.1 | - | 10 - 11 | 1224 | 94 | |
More Information
Set Membership: |
|

aclya
Genomic Map