Assay Details
| Gene Symbol: aclya | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002649.2 | NP_001002649.2 | 19 - 20 | 2466 | 67 | |
| GenBank mRNA | BC076484.1 | - | 19 - 20 | 2404 | 67 | |
More Information
Set Membership: |
|

aclya
Genomic Map