Assay Details
| Gene Symbol: serpinb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002653.1 | NP_001002653.1 | 5 - 6 | 624 | 111 | |
| GenBank mRNA | BC076524.1 | - | 5 - 6 | 624 | 111 | |
| BC133835.1 | - | 5 - 6 | 620 | 111 | ||
| BC165869.1 | - | 4 - 5 | 604 | 111 | ||
More Information
Set Membership: |
|

serpinb1
Genomic Map