Assay Details
| Gene Symbol: cox20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002712.2 | NP_001002712.2 | 2 - 3 | 283 | 67 | |
| GenBank mRNA | BC076091.1 | - | 2 - 3 | 195 | 67 | |
| EH537365.1 | - | 2 - 3 | 207 | 67 | ||
| EH553945.1 | - | 2 - 3 | 318 | 67 | ||
More Information
Set Membership: |
|

cox20
Genomic Map