Assay Details
| Gene Symbol: cox20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002712.2 | NP_001002712.2 | 3 - 4 | 340 | 71 | |
| GenBank mRNA | BC076091.1 | - | 3 - 4 | 252 | 71 | |
| EH537365.1 | - | 3 - 4 | 264 | 71 | ||
| EH553945.1 | - | 3 - 4 | 375 | 71 | ||
More Information
Set Membership: |
|

cox20
Genomic Map