Assay Details
| Gene Symbol: glrx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005950.1 | NP_001005950.1 | 8 - 9 | 810 | 101 | |
| GenBank mRNA | BC083453.1 | - | 8 - 9 | 810 | 101 | |
| BC153570.1 | - | 8 - 9 | 871 | 101 | ||
| BC164130.1 | - | 8 - 9 | 831 | 101 | ||
More Information
Set Membership: |
|

glrx3
Genomic Map