Assay Details
| Gene Symbol: lrrc23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006054.1 | NP_001006054.1 | 3 - 4 | 754 | 62 | |
| GenBank mRNA | BC083278.1 | - | 3 - 4 | 754 | 62 | |
More Information
Set Membership: |
|

lrrc23
Genomic Map