Assay Details
| Gene Symbol: gpsm1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007778.1 | NP_001007779.1 | 10 - 11 | 1406 | 129 | |
| GenBank mRNA | AY619723.1 | - | 10 - 11 | 1406 | 129 | |
| DQ235093.1 | - | 10 - 11 | 1352 | 129 | ||
More Information
Set Membership: |
|

gpsm1b
Genomic Map