Assay Details
| Gene Symbol: pex19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017399.1 | NP_001017399.1 | 6 - 7 | 751 | 73 | |
| GenBank mRNA | BC056815.1 | - | 6 - 7 | 770 | 73 | |
| BC092974.1 | - | 6 - 7 | 751 | 73 | ||
More Information
Set Membership: |
|

pex19
Genomic Map