Assay Details
Gene Symbol: nmt1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020480.1 | NP_001018316.1 | 2 - 3 | 220 | 149 | |
GenBank mRNA | AB194986.1 | - | 2 - 3 | 235 | 149 | |
BC056805.1 | - | 2 - 3 | 149 | |||
BC095383.1 | - | 2 - 3 | 220 | 149 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
