Assay Details
Gene Symbol: nmt1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020480.1 | NP_001018316.1 | 6 - 7 | 702 | 77 | |
GenBank mRNA | AB194986.1 | - | 6 - 7 | 717 | 77 | |
BC056805.1 | - | 6 - 7 | 713 | 77 | ||
BC095383.1 | - | 6 - 7 | 702 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
