Assay Details
| Gene Symbol: alkbh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020691.1 | NP_001018527.1 | 1 - 2 | 131 | 84 | |
| GenBank mRNA | BC095784.1 | - | 1 - 2 | 131 | 84 | |
| BC164762.1 | - | 1 - 2 | 155 | 84 | ||
More Information
Set Membership: |
|

alkbh1
Genomic Map