Assay Details
| Gene Symbol: tnfb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024447.1 | NP_001019618.1 | 1 - 2 | 217 | 69 | |
| GenBank mRNA | AB183467.1 | - | 1 - 2 | 217 | 69 | |
| BC116495.1 | - | 1 - 2 | 227 | 69 | ||
| BC164826.1 | - | 1 - 2 | 209 | 69 | ||
More Information
Set Membership: |
|

tnfb
Genomic Map