Assay Details
| Gene Symbol: tnfb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024447.1 | NP_001019618.1 | 3 - 4 | 355 | 64 | |
| GenBank mRNA | AB183467.1 | - | 3 - 4 | 355 | 64 | |
| BC116495.1 | - | 3 - 4 | 365 | 64 | ||
| BC164826.1 | - | 3 - 4 | 347 | 64 | ||
More Information
Set Membership: |
|

tnfb
Genomic Map