Assay Details
| Gene Symbol: rnf34a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030068.1 | NP_001025239.1 | 2 - 3 | 314 | 82 | |
| XM_005172464.2 | XP_005172521.1 | 2 - 3 | 382 | 82 | ||
| GenBank mRNA | BC097108.1 | - | 2 - 3 | 314 | 82 | |
More Information
Set Membership: |
|

rnf34a
Genomic Map