Assay Details
| Gene Symbol: srsf7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030267.1 | NP_001025438.1 | 286 | 148 | ||
| GenBank mRNA | BC097250.1 | - | 286 | 148 | ||
| BC133851.1 | - | 2 - 3 | 489 | 148 | ||
More Information
Set Membership: |
|

srsf7b
Genomic Map