Assay Details
| Gene Symbol: eif3hb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035264.2 | NP_001030341.1 | 4 - 5 | 551 | 90 | |
| GenBank mRNA | BC086809.1 | - | 4 - 5 | 517 | 90 | |
| BC107824.1 | - | 4 - 5 | 552 | 90 | ||
More Information
Set Membership: |
|

eif3hb
Genomic Map