Assay Details
| Gene Symbol: bmpr2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039807.1 | NP_001034896.1 | 11 - 12 | 2397 | 133 | |
| GenBank mRNA | BC163200.1 | - | 11 - 12 | 2255 | 133 | |
| DQ002899.1 | - | 11 - 12 | 2397 | 133 | ||
More Information
Set Membership: |
|

bmpr2b
Genomic Map