Assay Details
| Gene Symbol: slc43a3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039922.1 | NP_001035011.1 | 11 - 12 | 1502 | 90 | |
| XM_009291002.3 | XP_009289277.1 | 12 - 13 | 1639 | 90 | ||
| GenBank mRNA | BC114300.1 | - | 11 - 12 | 1502 | 90 | |
More Information
Set Membership: |
|

slc43a3b
Genomic Map