Assay Details
Gene Symbol: tfe3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045066.1 | NP_001038531.1 | 1 - 2 | 198 | 71 | |
XM_005169305.4 | XP_005169362.1 | 2 - 3 | 316 | 71 | ||
XM_005169306.4 | XP_005169363.1 | 2 - 3 | 316 | 71 | ||
XM_005169307.4 | XP_005169364.1 | 2 - 3 | 316 | 71 | ||
XM_005169308.4 | XP_005169365.1 | 2 - 3 | 316 | 71 | ||
XM_009306036.3 | XP_009304311.1 | 2 - 3 | 316 | 71 | ||
GenBank mRNA | JN105115.1 | - | 1 - 2 | 207 | 71 | |
JN105116.1 | - | 1 - 2 | 30 | 71 | ||
JN105117.1 | - | 1 - 2 | 198 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
