Assay Details
| Gene Symbol: pter | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045381.2 | NP_001038846.2 | 3 - 4 | 763 | 65 | |
| GenBank mRNA | BC122370.1 | - | 3 - 4 | 750 | 65 | |
| EH450120.1 | - | 3 - 4 | 789 | 65 | ||
| EH572300.1 | - | 2 - 3 | 389 | 65 | ||
More Information
Set Membership: |
|

pter
Genomic Map