Assay Details
| Gene Symbol: dhrs12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076557.1 | NP_001070025.1 | 8 - 9 | 715 | 75 | |
| GenBank mRNA | BC122377.1 | - | 8 - 9 | 715 | 75 | |
More Information
Set Membership: |
|

dhrs12
Genomic Map