Assay Details
Gene Symbol: ddx28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077332.1 | NP_001070800.1 | 1 | 1647 | 119 | |
GenBank mRNA | BC124668.1 | - | 1 | 1647 | 119 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
