Assay Details
| Gene Symbol: crtc1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077457.1 | NP_001070925.1 | 7 - 8 | 701 | 85 | |
| XM_009299199.3 | XP_009297474.1 | 7 - 8 | 1035 | 85 | ||
| XM_009299205.3 | XP_009297480.1 | 7 - 8 | 1024 | 85 | ||
| XM_021481340.1 | XP_021337015.1 | 7 - 8 | 1037 | 85 | ||
| GenBank mRNA | BC124409.1 | - | 7 - 8 | 701 | 85 | |
| BC165399.1 | - | 7 - 8 | 701 | 85 | ||
More Information
Set Membership: |
|

crtc1a
Genomic Map