Assay Details
Gene Symbol: tinf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005172300.4 | XP_005172357.1 | 5 - 6 | 1002 | 89 | |
XM_009303073.3 | XP_009301348.1 | 6 - 7 | 652 | 89 | ||
GenBank mRNA | BC128834.1 | - | 5 - 6 | 773 | 89 | |
EH501901.1 | - | 5 - 6 | 868 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
