Assay Details
| Gene Symbol: gnb5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080181.1 | NP_001073650.1 | 10 - 11 | 1159 | 141 | |
| GenBank mRNA | BC129375.1 | - | 10 - 11 | 1159 | 141 | |
More Information
Set Membership: |
|

gnb5a
Genomic Map