Assay Details
Gene Symbol: dtnbb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080573.1 | NP_001074042.1 | 9 - 10 | 1093 | 129 | |
XM_017351659.2 | XP_017207148.1 | 11 - 12 | 1568 | 129 | ||
XM_021467184.1 | XP_021322859.1 | 12 - 13 | 1649 | 129 | ||
XM_021467185.1 | XP_021322860.1 | 12 - 13 | 1649 | 129 | ||
XM_021467186.1 | XP_021322861.1 | 12 - 13 | 1650 | 129 | ||
XM_021467187.1 | XP_021322862.1 | 12 - 13 | 1649 | 129 | ||
GenBank mRNA | DQ516345.1 | - | 9 - 10 | 1093 | 129 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
