Assay Details
| Gene Symbol: pcyt1ba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082946.1 | NP_001076415.1 | 7 - 8 | 925 | 56 | |
| XM_005162665.4 | XP_005162722.1 | 7 - 8 | 1975 | 56 | ||
| XM_021470092.1 | XP_021325767.1 | 7 - 8 | 869 | 56 | ||
| GenBank mRNA | BC133902.1 | - | 7 - 8 | 925 | 56 | |
More Information
Set Membership: |
|

pcyt1ba
Genomic Map