Assay Details
| Gene Symbol: cacnb2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083568.1 | NP_001077037.1 | 8 - 9 | 758 | 73 | |
| GenBank mRNA | DQ372945.1 | - | 8 - 9 | 758 | 73 | |
| EU301434.1 | - | 8 - 9 | 799 | 73 | ||
| EU301435.1 | - | 8 - 9 | 685 | 73 | ||
| EU301436.1 | - | 9 - 10 | 819 | 73 | ||
| EU301437.1 | - | 9 - 10 | 1052 | 73 | ||
| EU301438.1 | - | 8 - 9 | 1076 | 73 | ||
| EU301439.1 | - | 8 - 9 | 962 | 73 | ||
| EU301440.1 | - | 9 - 10 | 1096 | 73 | ||
| EU301441.1 | - | 9 - 10 | 1329 | 73 | ||
More Information
Set Membership: |
|

cacnb2a
Genomic Map