Assay Details
| Gene Symbol: cacnb2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083568.1 | NP_001077037.1 | 9 - 10 | 864 | 65 | |
| GenBank mRNA | DQ372945.1 | - | 9 - 10 | 864 | 65 | |
| EU301434.1 | - | 9 - 10 | 905 | 65 | ||
| EU301435.1 | - | 9 - 10 | 791 | 65 | ||
| EU301436.1 | - | 10 - 11 | 925 | 65 | ||
| EU301437.1 | - | 10 - 11 | 1158 | 65 | ||
| EU301438.1 | - | 9 - 10 | 1182 | 65 | ||
| EU301439.1 | - | 9 - 10 | 1068 | 65 | ||
| EU301440.1 | - | 10 - 11 | 1202 | 65 | ||
| EU301441.1 | - | 10 - 11 | 1435 | 65 | ||
More Information
Set Membership: |
|

cacnb2a
Genomic Map