Assay Details
| Gene Symbol: cacnb2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083568.1 | NP_001077037.1 | 10 - 11 | 1013 | 76 | |
| GenBank mRNA | DQ372945.1 | - | 10 - 11 | 1013 | 76 | |
More Information
Set Membership: |
|

cacnb2a
Genomic Map