Assay Details
Gene Symbol: pip5k1ab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001089353.1 | NP_001082822.1 | 13 - 14 | 1616 | 126 | |
XM_021466679.1 | XP_021322354.1 | 14 - 15 | 1689 | 126 | ||
GenBank mRNA | BC129481.1 | - | 13 - 14 | 1616 | 126 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
