Assay Details
Gene Symbol: pip5k1ab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001089353.1 | NP_001082822.1 | 8 - 9 | 1096 | 75 | |
XM_021466679.1 | XP_021322354.1 | 9 - 10 | 1169 | 75 | ||
GenBank mRNA | BC129481.1 | - | 8 - 9 | 1096 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
