Assay Details
| Gene Symbol: pip5k1ab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001089353.1 | NP_001082822.1 | 9 - 10 | 1183 | 99 | |
| XM_021466679.1 | XP_021322354.1 | 10 - 11 | 1256 | 99 | ||
| GenBank mRNA | BC129481.1 | - | 9 - 10 | 1183 | 99 | |
More Information
Set Membership: |
|

pip5k1ab
Genomic Map