Assay Details
| Gene Symbol: nos2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104937.1 | NP_001098407.1 | 11 - 12 | 1398 | 95 | |
| XM_005165296.4 | XP_005165353.1 | 11 - 12 | 1466 | 95 | ||
| GenBank mRNA | AM084880.1 | - | 11 - 12 | 1398 | 95 | |
| AM749801.1 | - | 11 - 12 | 1398 | 95 | ||
| AY324390.1 | - | 1 - 2 | 53 | 95 | ||
More Information
Set Membership: |
|

nos2a
Genomic Map