Assay Details
Gene Symbol: nos2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104937.1 | NP_001098407.1 | 18 - 19 | 2151 | 83 | |
XM_005165296.4 | XP_005165353.1 | 18 - 19 | 2219 | 83 | ||
GenBank mRNA | AM084880.1 | - | 18 - 19 | 2151 | 83 | |
AM749801.1 | - | 18 - 19 | 2151 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
