Assay Details
| Gene Symbol: lipea | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001316725.1 | NP_001303654.1 | 10 - 11 | 2094 | 124 | |
| XM_005159495.4 | XP_005159552.1 | 10 - 11 | 2119 | 124 | ||
| GenBank mRNA | BC151926.1 | - | 10 - 11 | 2094 | 124 | |
More Information
Set Membership: |
|

lipea
Genomic Map