Assay Details
| Gene Symbol: khdrbs1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001362251.1 | NP_001349180.1 | 1 - 2 | 186 | 128 | |
| NM_001362252.1 | NP_001349181.1 | 1 - 2 | 186 | 128 | ||
| NM_130925.2 | NP_571000.1 | 1 - 2 | 186 | 128 | ||
| NR_155504.1 | 1 - 2 | 186 | 128 | |||
| NR_155505.1 | 1 - 2 | 186 | 128 | |||
| NR_155506.1 | 1 - 2 | 186 | 128 | |||
| NR_155507.1 | 1 - 2 | 186 | 128 | |||
| NR_155508.1 | 1 - 2 | 186 | 128 | |||
| NR_155509.1 | 1 - 2 | 186 | 128 | |||
| NR_155510.1 | 1 - 2 | 186 | 128 | |||
| NR_155512.1 | 1 - 2 | 186 | 128 | |||
| XM_009307570.3 | XP_009305845.1 | 1 - 2 | 269 | 128 | ||
| XM_017358677.2 | XP_017214166.1 | 1 - 2 | 269 | 128 | ||
| GenBank mRNA | BC067717.1 | - | 1 - 2 | 186 | 128 | |
| BC153455.1 | - | 1 - 2 | 197 | 128 | ||
More Information
Set Membership: |
|

khdrbs1a
Genomic Map